Learn More
Hach Company SC4500 Ctrl C1D2 Prog 5x mA Ou
Catalog No. 5000207348 Shop All Hach Company Products
Encompass
SC4500 Controller-C1D2-Prognosys-5x mA Output-1 digital Sensor-24 VDC-without plugIncludes SC4500 Controller-Claros-enabled-with EU plug includes mounting hardware
Catalog No. 50-002-07348 Supplier Hach Company Supplier No. LXV525.99Y11501
SC4500 Controller-C1D2-Prognosys-5x mA Output-1 digital Sensor-24 VDC-without plugIncludes SC4500 Controller-Claros-enabled-with EU plug includes mounting hardware