Learn More
Hach Company SC4500 Ctrl C1D2 Prog LAN mA O
Catalog No. 5000207666 Shop All Hach Company Products
Encompass
SC4500 Controller-C1D2-Prognosys-LAN mA Output-2 mA Inputs-24 VDC-without plugIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware
Catalog No. 50-002-07666 Supplier Hach Company Supplier No. LXV525.99Y1B441
SC4500 Controller-C1D2-Prognosys-LAN mA Output-2 mA Inputs-24 VDC-without plugIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware