Learn More
Hach Company SC4500 Ctrl C1D2 Prog Prof DP
Catalog No. 5000207458 Shop All Hach Company Products
Encompass
SC4500 Controller-C1D2-Prognosys-Profibus DP-1 digital Sensor-24 VDC-without plugIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware
Catalog No. 50-002-07458 Supplier Hach Company Supplier No. LXV525.99Y12501
SC4500 Controller-C1D2-Prognosys-Profibus DP-1 digital Sensor-24 VDC-without plugIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware