Learn More
Hach Company SC4500 Ctrl C1D2 Prog Prof DP
Catalog No. 5000208109 Shop All Hach Company Products
Encompass
SC4500 Controller-C1D2-Prognosys-Profibus DP-2 digital Sensors-24 VDC-without plugIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware
Catalog No. 50-002-08109 Supplier Hach Company Supplier No. LXV525.99Y12551
SC4500 Controller-C1D2-Prognosys-Profibus DP-2 digital Sensors-24 VDC-without plugIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware