Learn More
Hach Company SC4500 Ctrl Claros 5x mA Out 1
Catalog No. 5000207469 Shop All Hach Company Products
Encompass
SC4500 Controller-Claros-enabled-5x mA Output-1 digital Sensor-24 VDC-without plugIncludes SC4500 Controller-Claros-enabled-with EU plug includes mounting hardware
Catalog No. 50-002-07469 Supplier Hach Company Supplier No. LXV525.99ZA1501
SC4500 Controller-Claros-enabled-5x mA Output-1 digital Sensor-24 VDC-without plugIncludes SC4500 Controller-Claros-enabled-with EU plug includes mounting hardware