Learn More
Hach Company SC4500 Ctrl Claros 5x mA Out 1
Catalog No. 5000207908 Shop All Hach Company Products
Encompass
SC4500 Controller-Claros-enabled-5x mA Output-1 digital Sensor-1 analog pH/ORP-100-240 VAC-without power cordIncludes SC4500 Controller-Claros-enabled-with EU plug includes mounting hardware
Catalog No. 50-002-07908 Supplier Hach Company Supplier No. LXV525.99AA1511
SC4500 Controller-Claros-enabled-5x mA Output-1 digital Sensor-1 analog pH/ORP-100-240 VAC-without power cordIncludes SC4500 Controller-Claros-enabled-with EU plug includes mounting hardware