Learn More
Hach Company SC4500 Ctrl Claros 5x mA Out 1
Catalog No. 5000207818 Shop All Hach Company Products
Encompass
SC4500 Controller-Claros-enabled-5x mA Output-1 digital Sensor-1 analog Conductivity-100-240 VAC-EU plugIncludes SC4500 Controller-Claros-enabled-with EU plug includes mounting hardware
Catalog No. 50-002-07818 Supplier Hach Company Supplier No. LXV525.99CA1521
SC4500 Controller-Claros-enabled-5x mA Output-1 digital Sensor-1 analog Conductivity-100-240 VAC-EU plugIncludes SC4500 Controller-Claros-enabled-with EU plug includes mounting hardware