Learn More
Hach Company SC4500 Ctrl Claros 5x mA Out 2
Catalog No. 5000207669 Shop All Hach Company Products
Encompass
SC4500 Controller-Claros-enabled-5x mA Output-2 analog Conductivity-100-240 VAC-without power cordIncludes SC4500 Controller-Claros-enabled-with EU plug includes mounting hardware
Catalog No. 50-002-07669 Supplier Hach Company Supplier No. LXV525.99AA1221
SC4500 Controller-Claros-enabled-5x mA Output-2 analog Conductivity-100-240 VAC-without power cordIncludes SC4500 Controller-Claros-enabled-with EU plug includes mounting hardware