Learn More
Hach Company SC4500 Ctrl Claros LAN EthIP 2
Catalog No. 5000207303 Shop All Hach Company Products
Encompass
SC4500 Controller-Claros-enabled-LAN Ethernet IP-2 digital Sensors-100-240 VAC-US plugIncludes SC4500 Controller-Claros-enabled-with EU plug includes mounting hardware
Catalog No. 50-002-07303 Supplier Hach Company Supplier No. LXV525.99EAG551
SC4500 Controller-Claros-enabled-LAN Ethernet IP-2 digital Sensors-100-240 VAC-US plugIncludes SC4500 Controller-Claros-enabled-with EU plug includes mounting hardware