Learn More
Hach Company SC4500 Ctrl Claros LAN mA Out
Catalog No. 5000207561 Shop All Hach Company Products
Encompass
SC4500 Controller-Claros-enabled-LAN mA Output-1 digital Sensor 1 analog UPW Conductivity Sensor-100-240 VAC-EU plugIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware
Catalog No. 50-002-07561 Supplier Hach Company Supplier No. LXV525.99CAB561
SC4500 Controller-Claros-enabled-LAN mA Output-1 digital Sensor 1 analog UPW Conductivity Sensor-100-240 VAC-EU plugIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware