Learn More
Hach Company SC4500 Ctrl Claros LAN mA Out
Catalog No. 5000207302 Shop All Hach Company Products
Encompass
SC4500 Controller-Claros-enabled-LAN mA Output-1 digital Sensor-1 mA Input-100-240 VAC-US plugIncludes SC4500 Controller-Claros-enabled-with EU plug includes mounting hardware
Catalog No. 50-002-07302 Supplier Hach Company Supplier No. LXV525.99EAB541
SC4500 Controller-Claros-enabled-LAN mA Output-1 digital Sensor-1 mA Input-100-240 VAC-US plugIncludes SC4500 Controller-Claros-enabled-with EU plug includes mounting hardware