Learn More
Hach Company SC4500 Ctrl Claros LAN mA Out
Catalog No. 5000207910 Shop All Hach Company Products
Encompass
SC4500 Controller-Claros-enabled-LAN mA Output-1 Analog UPW pH/ORP Sensor-100-240 VAC-without power cordIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware
Catalog No. 50-002-07910 Supplier Hach Company Supplier No. LXV525.99AAB701
SC4500 Controller-Claros-enabled-LAN mA Output-1 Analog UPW pH/ORP Sensor-100-240 VAC-without power cordIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware