Learn More
Hach Company SC4500 Ctrl Claros LAN mA Out
Catalog No. 5000208010 Shop All Hach Company Products
Encompass
SC4500 Controller-Claros-enabled-LAN mA Output-2 digital Sensors-100-240 VAC-without power cordIncludes SC4500 Controller-Claros-enabled-with EU plug includes mounting hardware
Catalog No. 50-002-08010 Supplier Hach Company Supplier No. LXV525.99AAB551
SC4500 Controller-Claros-enabled-LAN mA Output-2 digital Sensors-100-240 VAC-without power cordIncludes SC4500 Controller-Claros-enabled-with EU plug includes mounting hardware