Learn More
Hach Company SC4500 Ctrl Claros LAN Modb RS
Catalog No. 5000207917 Shop All Hach Company Products
Encompass
SC4500 Controller-Claros-enabled-LAN Modbus RS-1 digital Sensor-24 VDC-without power cordIncludes SC4500 Controller-Claros-enabled-with EU plug includes mounting hardware
Catalog No. 50-002-07917 Supplier Hach Company Supplier No. LXV525.99ZAD501
SC4500 Controller-Claros-enabled-LAN Modbus RS-1 digital Sensor-24 VDC-without power cordIncludes SC4500 Controller-Claros-enabled-with EU plug includes mounting hardware