Learn More
Hach Company SC4500 Ctrl Claros LAN Prof DP
Catalog No. 5000207815 Shop All Hach Company Products
Encompass
SC4500 Controller-Claros-enabled-LAN Profibus DP-2 Analog UPW pH/ORP Sensors-100-240 VAC-without power cordIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware
Catalog No. 50-002-07815 Supplier Hach Company Supplier No. LXV525.99AAC771
SC4500 Controller-Claros-enabled-LAN Profibus DP-2 Analog UPW pH/ORP Sensors-100-240 VAC-without power cordIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware