Learn More
Hach Company SC4500 Ctrl Claros LAN Profi 2
Catalog No. 5000207660 Shop All Hach Company Products
Encompass
SC4500 Controller-Claros-enabled-LAN Profinet-2 digital Sensors-100-240 VAC-without power cordIncludes SC4500 Controller-Claros-enabled-with EU plug includes mounting hardware
Catalog No. 50-002-07660 Supplier Hach Company Supplier No. LXV525.99AAF551
SC4500 Controller-Claros-enabled-LAN Profinet-2 digital Sensors-100-240 VAC-without power cordIncludes SC4500 Controller-Claros-enabled-with EU plug includes mounting hardware