Learn More
Hach Company SC4500 Ctrl Claros Modb TCP LA
Catalog No. 5000207338 Shop All Hach Company Products
Encompass
SC4500 Controller-Claros-enabled-Modbus TCP LAN-2 digital Sensors-24 VDC-without plugIncludes SC4500 Controller-Claros-enabled-with EU plug includes mounting hardware
Catalog No. 50-002-07338 Supplier Hach Company Supplier No. LXV525.99ZA5551
SC4500 Controller-Claros-enabled-Modbus TCP LAN-2 digital Sensors-24 VDC-without plugIncludes SC4500 Controller-Claros-enabled-with EU plug includes mounting hardware