Learn More
Hach Company SC4500 Ctrl Claros Prof DP 2 d
Catalog No. 5000207336 Shop All Hach Company Products
Encompass
SC4500 Controller-Claros-enabled-Profibus DP-2 digital Sensors-100-240 VAC-without power cordIncludes SC4500 Controller-Claros-enabled-with EU plug includes mounting hardware
Catalog No. 50-002-07336 Supplier Hach Company Supplier No. LXV525.99AA2551
SC4500 Controller-Claros-enabled-Profibus DP-2 digital Sensors-100-240 VAC-without power cordIncludes SC4500 Controller-Claros-enabled-with EU plug includes mounting hardware