Learn More
Hach Company SC4500 Ctrl LAN EthIP 2 mA Ins
Catalog No. 5000207568 Shop All Hach Company Products
Encompass
SC4500 Controller-LAN Ethernet IP-2 mA Inputs-24 VDC-without plugIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware
Catalog No. 50-002-07568 Supplier Hach Company Supplier No. LXV525.99Z0G441
SC4500 Controller-LAN Ethernet IP-2 mA Inputs-24 VDC-without plugIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware