Learn More
Hach Company SC4500 Ctrl Modb TCP LAN 1 dig
Catalog No. 5000207400 Shop All Hach Company Products
Encompass
SC4500 Controller-Modbus TCP LAN-1 digital Sensor-1 mA Input-24 VDC-without plugIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware
Catalog No. 50-002-07400 Supplier Hach Company Supplier No. LXV525.99Z05541
SC4500 Controller-Modbus TCP LAN-1 digital Sensor-1 mA Input-24 VDC-without plugIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware