Learn More
Hach Company SC4500 Ctrl Prog 5x mA Out 1 a
Catalog No. 5000207763 Shop All Hach Company Products
Encompass
SC4500 Controller-Prognosys-5x mA Output-1 analog Conductivity-100-240 VAC-without plug C1D2Includes SC4500 Controller-Prognosys-without plug C1D2 includes mounting hardware
Catalog No. 50-002-07763 Supplier Hach Company Supplier No. LXV525.99P11201
SC4500 Controller-Prognosys-5x mA Output-1 analog Conductivity-100-240 VAC-without plug C1D2Includes SC4500 Controller-Prognosys-without plug C1D2 includes mounting hardware