Learn More
Hach Company SC4500 Ctrl Prog 5x mA Out 1 a
Catalog No. 5000207467 Shop All Hach Company Products
Encompass
SC4500 Controller-Prognosys-5x mA Output-1 analog pH/ORP-24 VDC-without plugIncludes SC4500 Controller-Claros-enabled-with EU plug includes mounting hardware
Catalog No. 50-002-07467 Supplier Hach Company Supplier No. LXV525.99Z11101
SC4500 Controller-Prognosys-5x mA Output-1 analog pH/ORP-24 VDC-without plugIncludes SC4500 Controller-Claros-enabled-with EU plug includes mounting hardware