Learn More
Hach Company SC4500 Ctrl Prog 5x mA Out 1 d
Catalog No. 5000207892 Shop All Hach Company Products
Encompass
SC4500 Controller-Prognosys-5x mA Output-1 digital Sensor-1 analog Conductivity-100-240 VAC-without power cordIncludes SC4500 Controller-Claros-enabled-with EU plug includes mounting hardware
Catalog No. 50-002-07892 Supplier Hach Company Supplier No. LXV525.99A11521
SC4500 Controller-Prognosys-5x mA Output-1 digital Sensor-1 analog Conductivity-100-240 VAC-without power cordIncludes SC4500 Controller-Claros-enabled-with EU plug includes mounting hardware