Learn More
Hach Company SC4500 Ctrl Prog 5x mA Out 1 m
Catalog No. 5000207948 Shop All Hach Company Products
Encompass
SC4500 Controller-Prognosys-5x mA Output-1 mA Input-100-240 VAC-without power cordIncludes SC4500 Controller-Claros-enabled-with EU plug includes mounting hardware
Catalog No. 50-002-07948 Supplier Hach Company Supplier No. LXV525.99A11401
SC4500 Controller-Prognosys-5x mA Output-1 mA Input-100-240 VAC-without power cordIncludes SC4500 Controller-Claros-enabled-with EU plug includes mounting hardware