Learn More
Hach Company SC4500 Ctrl Prog 5x mA Out 2 m
Catalog No. 5000207829 Shop All Hach Company Products
Encompass
SC4500 Controller-Prognosys-5x mA Output-2 mA Inputs-100-240 VAC-without plug C1D2Includes SC4500 Controller-Claros-enabled-with EU plug includes mounting hardware
Catalog No. 50-002-07829 Supplier Hach Company Supplier No. LXV525.99P11441
SC4500 Controller-Prognosys-5x mA Output-2 mA Inputs-100-240 VAC-without plug C1D2Includes SC4500 Controller-Claros-enabled-with EU plug includes mounting hardware