Learn More
Hach Company SC4500 Ctrl Prog EthIP 2 mA In
Catalog No. 5000208011 Shop All Hach Company Products
Encompass
SC4500 Controller-Prognosys-Ethernet IP-2 mA Inputs-24 VDC-without plugIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware
Catalog No. 50-002-08011 Supplier Hach Company Supplier No. LXV525.99Z17441
SC4500 Controller-Prognosys-Ethernet IP-2 mA Inputs-24 VDC-without plugIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware