Learn More
Hach Company SC4500 Ctrl Prog LAN EthIP 1 d
Catalog No. 5000207569 Shop All Hach Company Products
Encompass
SC4500 Controller-Prognosys-LAN Ethernet IP-1 digital Sensor-1 mA Input-100-240 VAC-without power cordIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware
Catalog No. 50-002-07569 Supplier Hach Company Supplier No. LXV525.99A1G541
SC4500 Controller-Prognosys-LAN Ethernet IP-1 digital Sensor-1 mA Input-100-240 VAC-without power cordIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware