Learn More
Hach Company SC4500 Ctrl Prog LAN EthIP 2 d
Catalog No. 5000207298 Shop All Hach Company Products
Encompass
SC4500 Controller-Prognosys-LAN Ethernet IP-2 digital Sensors 100-240 VAC-US plugIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware
Catalog No. 50-002-07298 Supplier Hach Company Supplier No. LXV525.99E1G551
SC4500 Controller-Prognosys-LAN Ethernet IP-2 digital Sensors 100-240 VAC-US plugIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware