Learn More
Hach Company SC4500 Ctrl Prog LAN mA Out 1
Catalog No. 5000207368 Shop All Hach Company Products
Encompass
SC4500 Controller-Prognosys-LAN mA Output-1 digital Sensor-1 mA Input-100-240 VAC-without plug C1D2Includes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware
Catalog No. 50-002-07368 Supplier Hach Company Supplier No. LXV525.99P1B541
SC4500 Controller-Prognosys-LAN mA Output-1 digital Sensor-1 mA Input-100-240 VAC-without plug C1D2Includes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware