Learn More
Hach Company SC4500 Ctrl Prog LAN mA Out 2
Catalog No. 5000208050 Shop All Hach Company Products
Encompass
SC4500 Controller-Prognosys-LAN mA Output-2 mA Inputs-100-240 VAC-without plug C1D2Includes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware
Catalog No. 50-002-08050 Supplier Hach Company Supplier No. LXV525.99P1B441
SC4500 Controller-Prognosys-LAN mA Output-2 mA Inputs-100-240 VAC-without plug C1D2Includes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware