Learn More
Hach Company SC4500 Ctrl Prog LAN mA Out 2
Catalog No. 5000207306 Shop All Hach Company Products
Encompass
SC4500 Controller-Prognosys-LAN mA Output-2 Analog UPW Conductivity Sensors-100-240 VAC-without power cordIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware
Catalog No. 50-002-07306 Supplier Hach Company Supplier No. LXV525.99A1B661
SC4500 Controller-Prognosys-LAN mA Output-2 Analog UPW Conductivity Sensors-100-240 VAC-without power cordIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware