Learn More
Hach Company SC4500 Ctrl Prog LAN Modb RS 2
Catalog No. 5000207574 Shop All Hach Company Products
Encompass
SC4500 Controller-Prognosys-LAN Modbus RS-2 digital Sensors-24 VDC-without power cordIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware
Catalog No. 50-002-07574 Supplier Hach Company Supplier No. LXV525.99Z1D551
SC4500 Controller-Prognosys-LAN Modbus RS-2 digital Sensors-24 VDC-without power cordIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware