Learn More
Hach Company SC4500 Ctrl Prog LAN Prof 1 an
Catalog No. 5000207381 Shop All Hach Company Products
Encompass
SC4500 Controller-Prognosys-LAN Profibus-1 analog Conductivity-1 analog pH/ORP-100-240 VAC-EU plugIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware
Catalog No. 50-002-07381 Supplier Hach Company Supplier No. LXV525.99C1C211
SC4500 Controller-Prognosys-LAN Profibus-1 analog Conductivity-1 analog pH/ORP-100-240 VAC-EU plugIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware