Learn More
Hach Company SC4500 Ctrl Prog LAN Profi 2 m
Catalog No. 5000207345 Shop All Hach Company Products
Encompass
SC4500 Controller-Prognosys-LAN Profinet-2 mA Inputs-100-240 VAC-US plugIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware
Catalog No. 50-002-07345 Supplier Hach Company Supplier No. LXV525.99E1F441
SC4500 Controller-Prognosys-LAN Profinet-2 mA Inputs-100-240 VAC-US plugIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware