Learn More
Hach Company SC4500 Ctrl Prog mA Out 1 Anal
Catalog No. 5000208121 Shop All Hach Company Products
Encompass
SC4500 Controller-Prognosys-mA Output-1 Analog UPW pH/ORP Sensor-24 VDC-without power cordIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware
Catalog No. 50-002-08121 Supplier Hach Company Supplier No. LXV525.99Z11701
SC4500 Controller-Prognosys-mA Output-1 Analog UPW pH/ORP Sensor-24 VDC-without power cordIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware