Learn More
Hach Company SC4500 Ctrl Prog mA Out 1 Anal
Catalog No. 5000208027 Shop All Hach Company Products
Encompass
SC4500 Controller-Prognosys-mA Output-1 Analog UPW pH/ORP 1 Analog UPW Conductivity Sensor-100-240 VAC-without power cordIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware
Catalog No. 50-002-08027 Supplier Hach Company Supplier No. LXV525.99A11761
SC4500 Controller-Prognosys-mA Output-1 Analog UPW pH/ORP 1 Analog UPW Conductivity Sensor-100-240 VAC-without power cordIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware