Learn More
Hach Company SC4500 Ctrl Prog Modb RS 1 anl
Catalog No. 5000207976 Shop All Hach Company Products
Encompass
SC4500 Controller-Prognosys-Modbus RS-1 analog UPW pH/ORP-1 analog UPW Conductivity Sensor-100-240 VAC-without power cordIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware
Catalog No. 50-002-07976 Supplier Hach Company Supplier No. LXV525.99A13761
SC4500 Controller-Prognosys-Modbus RS-1 analog UPW pH/ORP-1 analog UPW Conductivity Sensor-100-240 VAC-without power cordIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware