Learn More
Hach Company SC4500 Ctrl Prog Modb TCP LAN
Catalog No. 5000207563 Shop All Hach Company Products
Encompass
SC4500 Controller-Prognosys-Modbus TCP LAN-2 digital Sensors-100-240 VAC-US plugIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware
Catalog No. 50-002-07563 Supplier Hach Company Supplier No. LXV525.99E15551
SC4500 Controller-Prognosys-Modbus TCP LAN-2 digital Sensors-100-240 VAC-US plugIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware