Learn More
Hach Company SC4500 Ctrl Prog Prof DP 1 Ana
Catalog No. 5000207356 Shop All Hach Company Products
Encompass
SC4500 Controller-Prognosys-Profibus DP-1 Analog UPW pH/ORP 1 Analog UPW Conductivity Sensor-100-240 VAC-w/o power cordIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware
Catalog No. 50-002-07356 Supplier Hach Company Supplier No. LXV525.99A12761
SC4500 Controller-Prognosys-Profibus DP-1 Analog UPW pH/ORP 1 Analog UPW Conductivity Sensor-100-240 VAC-w/o power cordIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware