Learn More
Hach Company SC4500 Ctrl Prog Prof DP 1 Ana
Catalog No. 5000207947 Shop All Hach Company Products
Encompass
SC4500 Controller-Prognosys-Profibus DP-1 Analog UPW Conductivity Sensor-100-240 VAC-without power cordIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware
Catalog No. 50-002-07947 Supplier Hach Company Supplier No. LXV525.99A12601
SC4500 Controller-Prognosys-Profibus DP-1 Analog UPW Conductivity Sensor-100-240 VAC-without power cordIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware