Learn More
Hach Company SC4500 Ctrl Prog Prof DP 2 Ana
Catalog No. 5000208063 Shop All Hach Company Products
Encompass
SC4500 Controller-Prognosys-Profibus DP-2 Analog UPW Conductivity Sensors-100-240 VAC-without power cordIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware
Catalog No. 50-002-08063 Supplier Hach Company Supplier No. LXV525.99A12661
SC4500 Controller-Prognosys-Profibus DP-2 Analog UPW Conductivity Sensors-100-240 VAC-without power cordIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware