Learn More
Hach Company SC4500 Ctrl Prog Prof DP 2 dig
Catalog No. 5000207407 Shop All Hach Company Products
Encompass
SC4500 Controller-Prognosys-Profibus DP-2 digital Sensors-24 VDC-without plugIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware
Catalog No. 50-002-07407 Supplier Hach Company Supplier No. LXV525.99Z12551
SC4500 Controller-Prognosys-Profibus DP-2 digital Sensors-24 VDC-without plugIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware