Learn More
Hach Company SC4500 Ctrl Prog Profi 1 dig S
Catalog No. 5000207677 Shop All Hach Company Products
Encompass
SC4500 Controller-Prognosys-Profinet-1 digital Sensor-1 mA Input-24 VDC-without plugIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware
Catalog No. 50-002-07677 Supplier Hach Company Supplier No. LXV525.99Z16541
SC4500 Controller-Prognosys-Profinet-1 digital Sensor-1 mA Input-24 VDC-without plugIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware