Learn More
Hach Company SC4500 Ctrl Prog Profi 2 dig S
Catalog No. 5000207560 Shop All Hach Company Products
Encompass
SC4500 Controller-Prognosys-Profinet-2 digital Sensors-100-240 VAC-US plugIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware
Catalog No. 50-002-07560 Supplier Hach Company Supplier No. LXV525.99E16551
SC4500 Controller-Prognosys-Profinet-2 digital Sensors-100-240 VAC-US plugIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware