Learn More
Hach Company SC4500 CtrlC1D2 Prog 5x mA Out
Catalog No. 5000207344 Shop All Hach Company Products
Encompass
SC4500 Controller C1D2-Prognosys-5x mA Output-1 digital Sensor-1 mA Input-24 VDC-without plugIncludes SC4500 Controller-Claros-enabled-with EU plug includes mounting hardware
Catalog No. 50-002-07344 Supplier Hach Company Supplier No. LXV525.99Y11541
SC4500 Controller C1D2-Prognosys-5x mA Output-1 digital Sensor-1 mA Input-24 VDC-without plugIncludes SC4500 Controller-Claros-enabled-with EU plug includes mounting hardware