Learn More
Hach Company SC4500 Ethernet IP Upgrade Kit
Catalog No. 5000207116 Shop All Hach Company Products
Encompass
Upgrade kit to add Ethernet IP communication capability for SC4500 versions without Ethernet output
Catalog No. 50-002-07116 Supplier Hach Company Supplier No. LXZ525.99.C0002
Upgrade kit to add Ethernet IP communication capability for SC4500 versions without Ethernet output