Learn More
Hach Company SC4500 Sunroof with visor
Catalog No. 5000207064 Shop All Hach Company Products
Encompass
The sun roof protects the SC4500 controller from damaging UV rays Pair with the UV protection screen for ultimate protection against the elements
Catalog No. 50-002-07064 Supplier Hach Company Supplier No. LXZ524.99.00037
The sun roof protects the SC4500 controller from damaging UV rays Pair with the UV protection screen for ultimate protection against the elements