Learn More
Cole-Parmer Environmental Express SC490 Digestion Tubes, White Screw-Top Caps, PP, 100 mL; 225/Pk
Catalog No. NC2131537 Shop All Cole-Parmer Products
Encompass
Approved for hot plate digestions and certified for low metals background.
This item is not returnable. View return policy
Catalog No. NC2131537 Supplier Cole-Parmer Supplier No. 5320107
Approved for hot plate digestions and certified for low metals background.