Learn More
Hach Company SC4x00 Profibus DP Net Module
Catalog No. 5000207109 Shop All Hach Company Products
Encompass
SC4x00 Profibus DP Network ModuleIncludes The Profibus DP communication module for the SC4200c is a user installed module that allows for communication via Profibus from the controller See manual for additional connection information
Catalog No. 50-002-07109 Supplier Hach Company Supplier No. LXZ524.99.00007
SC4x00 Profibus DP Network ModuleIncludes The Profibus DP communication module for the SC4200c is a user installed module that allows for communication via Profibus from the controller See manual for additional connection information