Learn More
Hach Company SC4x00 UV Protection Screen
Catalog No. 5000207079 Shop All Hach Company Products
Encompass
SC4x00 UV Protection Screen with SunroofIncludes This assembly covers the face of the SC4200c/SC4500 protecting it from the sun The transparent door can be opened for operating the controller It is UV absorbent and protects the controller from harmful UV rays Includes sunroof to further protect the controller
Catalog No. 50-002-07079 Supplier Hach Company Supplier No. LXZ524.99.00005
SC4x00 UV Protection Screen with SunroofIncludes This assembly covers the face of the SC4200c/SC4500 protecting it from the sun The transparent door can be opened for operating the controller It is UV absorbent and protects the controller from harmful UV rays Includes sunroof to further protect the controller